missing translation for 'onlineSavingsMsg'
Learn More

1,6-Bis(diphenylphosphino)hexane 95.0+%, TCI America™

Catalog No. B19591G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19591G 1 g
B19595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19591G Supplier TCI America Supplier No. B19591G

Chemical Identifiers

CAS 19845-69-3
Molecular Formula C30H32P2
Molecular Weight (g/mol) 454.53
MDL Number MFCD00003053
InChI Key GPORFKPYXATYNX-UHFFFAOYSA-N
Synonym 1,6-bis diphenylphosphino hexane, 6-diphenylphosphanylhexyl diphenyl phosphane, hexamethylenebis diphenylphosphine, phosphine, 1,6-hexanediylbis diphenyl, phosphine,1,1'-1,6-hexanediyl bis 1,1-diphenyl, 6-diphenylphosphanyl hexyl diphenylphosphane, acmc-1ca9e, ksc491k2l, 1,6-bis diphenylphosino hexane
PubChem CID 2754312
IUPAC Name [6-(diphenylphosphanyl)hexyl]diphenylphosphane
SMILES C(CCCP(C1=CC=CC=C1)C1=CC=CC=C1)CCP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 126°C
Color White-Yellow
Quantity 1 g
Formula Weight 454.53
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,6-Bis(diphenylphosphino)hexane