Learn More
Thermo Scientific Chemicals 1,6-Anhydro-beta-D-cellobiose, 95%
CAS: 35405-71-1 | C12H20O10 | 324.282 g/mol
Supplier: Thermo Scientific Chemicals J6691403
| Quantity | 1 g |
|---|
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 35405-71-1 | |
| 324.282 | |
| LTYZUJSCZCPGHH-QRZGKKJRSA-N | |
| 71225847 | |
| C1C2C(C(C(C(O1)O2)O)O)OC3C(C(C(C(O3)CO)O)O)O |
| C12H20O10 | |
| MFCD13182343 | |
| 1,6-anhydro-b-d-cellobiose, cellobiosan, 1,6-anhydro-beta-d-cellobiose, 2s,3r,4s,5s,6r-2-1r,2s,3r,4r,5r-3,4-dihydroxy-6,8-dioxabicyclo 3.2.1 octan-2-yl oxy-6-hydroxymethyl oxane-3,4,5-triol | |
| (2S,3R,4S,5S,6R)-2-[[(1R,2S,3R,4R,5R)-3,4-dihydroxy-6,8-dioxabicyclo[3.2.1]octan-2-yl]oxy]-6-(hydroxymethyl)oxane-3,4,5-triol |
Specifications
| 1,6-Anhydro-beta-D-cellobiose | |
| White | |
| 1 g | |
| 1,6-anhydro-b-d-cellobiose, cellobiosan, 1,6-anhydro-beta-d-cellobiose, 2s,3r,4s,5s,6r-2-1r,2s,3r,4r,5r-3,4-dihydroxy-6,8-dioxabicyclo 3.2.1 octan-2-yl oxy-6-hydroxymethyl oxane-3,4,5-triol | |
| LTYZUJSCZCPGHH-QRZGKKJRSA-N | |
| (2S,3R,4S,5S,6R)-2-[[(1R,2S,3R,4R,5R)-3,4-dihydroxy-6,8-dioxabicyclo[3.2.1]octan-2-yl]oxy]-6-(hydroxymethyl)oxane-3,4,5-triol | |
| 71225847 | |
| 95% |
| 35405-71-1 | |
| C12H20O10 | |
| MFCD13182343 | |
| Soluble in water at 50mg/ml | |
| C1C2C(C(C(C(O1)O2)O)O)OC3C(C(C(C(O3)CO)O)O)O | |
| 324.282 | |
| 324.28 | |
| Powder |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
TSCA : No
Recommended Storage : Ambient temperatures