Learn More
1,5-Pentanedithiol, 96%
CAS: 928-98-3 | C5H12S2 | 136.27 g/mol
$161.00 - $161.00
Chemical Identifiers
| CAS | 928-98-3 |
|---|---|
| Molecular Formula | C5H12S2 |
| Molecular Weight (g/mol) | 136.27 |
| MDL Number | MFCD00004908 |
| InChI Key | KMTUBAIXCBHPIZ-UHFFFAOYSA-N |
| Synonym | 1,5-pentanedithiol, 1,5-dimercaptopentane, unii-r8rc3q7t1w, pentamethylene dimercaptan, r8rc3q7t1w, pentamethylenedithiol, 1,5-pentanodithiol, acmc-209rih, pentamethylenedimercaptane, 1,5-pentanedithiol 98+% |
| PubChem CID | 70236 |
| IUPAC Name | pentane-1,5-dithiol |
| SMILES | SCCCCCS |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2566514
|
Thermo Scientific Chemicals
B2566514 |
25 g |
Each for $161.00
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 928-98-3 | |
| 136.27 | |
| KMTUBAIXCBHPIZ-UHFFFAOYSA-N | |
| 70236 | |
| SCCCCCS |
| C5H12S2 | |
| MFCD00004908 | |
| 1,5-pentanedithiol, 1,5-dimercaptopentane, unii-r8rc3q7t1w, pentamethylene dimercaptan, r8rc3q7t1w, pentamethylenedithiol, 1,5-pentanodithiol, acmc-209rih, pentamethylenedimercaptane, 1,5-pentanedithiol 98+% | |
| pentane-1,5-dithiol |
Specifications
| 928-98-3 | |
| 1.016 | |
| 95°C (203°F) | |
| C5H12S2 | |
| MFCD00004908 | |
| UN3334 | |
| Air Sensitive | |
| KMTUBAIXCBHPIZ-UHFFFAOYSA-N | |
| pentane-1,5-dithiol | |
| 70236 | |
| 96% |
| -72°C | |
| 216°C to 218°C | |
| Unpleasant | |
| 1.5194 | |
| 25 g | |
| 1732335 | |
| 1,5-pentanedithiol, 1,5-dimercaptopentane, unii-r8rc3q7t1w, pentamethylene dimercaptan, r8rc3q7t1w, pentamethylenedithiol, 1,5-pentanodithiol, acmc-209rih, pentamethylenedimercaptane, 1,5-pentanedithiol 98+% | |
| SCCCCCS | |
| 136.27 | |
| 136.28 | |
| 1,5-Pentanedithiol |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 213-194-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only