missing translation for 'onlineSavingsMsg'
Learn More

1,5-Naphthyridine 98.0+%, TCI America™

Catalog No. N04011G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N04011G 1 g
N0401100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N04011G Supplier TCI America Supplier No. N04011G

Chemical Identifiers

CAS 254-79-5
Molecular Formula C8H6N2
Molecular Weight (g/mol) 130.15
MDL Number MFCD00039727
InChI Key VMLKTERJLVWEJJ-UHFFFAOYSA-N
Synonym 1,5-diazanaphthalene, 1,5-pyridopyridine, pyridino 3,2-b pyridine, pyridopyridine, 1,5-naphtyridine, 1,5 naphthyridine, zlchem 698, pubchem18072, acmc-209gka, pyrido 1,5-b pyridine
PubChem CID 136070
ChEBI CHEBI:36625
IUPAC Name 1,5-naphthyridine
SMILES C1=CC2=NC=CC=C2N=C1

Specifications

Melting Point 74°C
Color Red-White
Boiling Point 152°C
Quantity 1 g
Formula Weight 130.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Naphthyridine