missing translation for 'onlineSavingsMsg'
Learn More

1,5-Naphthyridine, 97+%

Catalog No. p-7055810
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64484MD 250 mg
AAH6448403 1 g
AAH6448406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH64484MD Supplier Thermo Scientific Chemicals Supplier No. H64484MD

CAS: 254-79-5 | C8H6N2 | 130.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 254-79-5
Molecular Formula C8H6N2
Molecular Weight (g/mol) 130.15
MDL Number MFCD00039727
InChI Key VMLKTERJLVWEJJ-UHFFFAOYSA-N
Synonym 1,5-diazanaphthalene, 1,5-pyridopyridine, pyridino 3,2-b pyridine, pyridopyridine, 1,5-naphtyridine, 1,5 naphthyridine, zlchem 698, pubchem18072, acmc-209gka, pyrido 1,5-b pyridine
PubChem CID 136070
ChEBI CHEBI:36625
IUPAC Name 1,5-naphthyridine
SMILES C1=CC2=NC=CC=C2N=C1

Specifications

Melting Point 69°C to 75°C
Color White
Density 1.25 g/mL
Quantity 250 mg
Beilstein 109350
Formula Weight 130.15
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material 1,5-Naphthyridine
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only