missing translation for 'onlineSavingsMsg'
Learn More

1,5-Hexadiyne (stabilized with BHT) 95.0+%, TCI America™

Catalog No. H04855G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04855G 5 g
H04851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H04855G Supplier TCI America Supplier No. H04855G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 628-16-0
Molecular Formula C6H6
Molecular Weight (g/mol) 78.114
MDL Number MFCD00014924
InChI Key YFIBSNDOVCWPBL-UHFFFAOYSA-N
Synonym 1,5-hexadiyne, dipropargyl, bipropargyl, hc#cch2ch2c#ch, 1,5-hexadiyne in pentane, hccch2ch2cch, acmc-1b142, 1,5-hexadiyne 6ci, 8ci, 9ci, 1,5-hexadiyne solution, 50 wt. % in pentane 1g
PubChem CID 69402
ChEBI CHEBI:37821
IUPAC Name hexa-1,5-diyne
SMILES C#CCCC#C

Specifications

Color Yellow
Boiling Point 86°C
Quantity 5 g
UN Number 3295
Formula Weight 78.11
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,5-Hexadiyne (stabilized with BHT)
TSCA Yes
Recommended Storage Refrigerator