missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diphenyl-3-(2-thienyl)formazan 96.0+%, TCI America™

Catalog No. b0326100mg
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0326100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B0326100MG Supplier TCI America Supplier No. B0326100MG

Chemical Identifiers

CAS 135984-01-9
Molecular Formula C17H14N4S
Molecular Weight (g/mol) 306.387
MDL Number MFCD00059744
InChI Key YYNVZGCGVPELJN-GSPFKFNASA-N
Synonym STC Formazan
PubChem CID 66485618
IUPAC Name N'-anilino-N-phenyliminothiophene-2-carboximidamide
SMILES C1=CC=C(C=C1)NN=C(C2=CC=CS2)N=NC3=CC=CC=C3

Specifications

Melting Point 144°C
Color Black
Quantity 100 mg
Formula Weight 306.39
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Diphenyl-3-(2-thienyl)formazan