missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diphenyl-1,5-pentanedione, 99%

Catalog No. AC409010100
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC409010100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC409010100 Supplier Thermo Scientific Chemicals Supplier No. 409010100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6263-83-8 | C17H16O2 | 252.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6263-83-8
Molecular Formula C17H16O2
Molecular Weight (g/mol) 252.31
MDL Number MFCD00051401
InChI Key YOLLTWVIOASMFW-UHFFFAOYSA-N
Synonym 1,3-dibenzoylpropane, 1,5-diphenyl-1,5-pentanedione, 1,5-pentanedione, 1,5-diphenyl, 1,5-diphenyl-1,5-pentadione, 1,5-pentadione, maybridge3_005349, acmc-1b50l, 1,5-diphenyl-1,5-pentandione, 1,5-diphenyl-pentane-1,5-dione
PubChem CID 80432
IUPAC Name 1,5-diphenylpentane-1,5-dione
SMILES C1=CC=C(C=C1)C(=O)CCCC(=O)C2=CC=CC=C2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 64°C to 67°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 10 g
Formula Weight 252.31
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 1, 5-Diphenyl-1, 5-pentanedione, 99%
Show More Show Less

RUO – Research Use Only