missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dimethylnaphthalene 97.0+%, TCI America™

Supplier:  TCI America D27645G

 View more versions of this product

Catalog No. D27645G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1,5-Dimethylnaphthalene
81°C
C12H12
5 g
SDDBCEWUYXVGCQ-UHFFFAOYSA-N
1,5-dimethylnaphthalene
11306
156.23
Crystalline Powder
571-61-9
White-Yellow
MFCD00004038
naphthalene, 1,5-dimethyl, 1,5-dmn, 1,5-dimethyl-naphthalene, acmc-209lwa, naphthalene,5-dimethyl, 1,5-dimethyl naphthalene, 1,5-dimethylnaphthalene, naphthalene, 1,5-dimethyl-8ci 9ci
CC1=CC=CC2=C(C)C=CC=C12
156.23
CHEBI:48608
≥97.0% (GC)

Chemical Identifiers

571-61-9
156.23
SDDBCEWUYXVGCQ-UHFFFAOYSA-N
11306
1,5-dimethylnaphthalene
C12H12
MFCD00004038
naphthalene, 1,5-dimethyl, 1,5-dmn, 1,5-dimethyl-naphthalene, acmc-209lwa, naphthalene,5-dimethyl, 1,5-dimethyl naphthalene, 1,5-dimethylnaphthalene, naphthalene, 1,5-dimethyl-8ci 9ci
CHEBI:48608
CC1=CC=CC2=C(C)C=CC=C12
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0080

TSCA : No

SDS
Documents

Documents

Promotions

Promotions