missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dimethylnaphthalene 97.0+%, TCI America™

Catalog No. D27641G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27641G 1 g
D27645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27641G Supplier TCI America Supplier No. D27641G

Chemical Identifiers

CAS 571-61-9
Molecular Formula C12H12
Molecular Weight (g/mol) 156.23
MDL Number MFCD00004038
InChI Key SDDBCEWUYXVGCQ-UHFFFAOYSA-N
Synonym naphthalene, 1,5-dimethyl, 1,5-dmn, 1,5-dimethyl-naphthalene, acmc-209lwa, naphthalene,5-dimethyl, 1,5-dimethyl naphthalene, 1,5-dimethylnaphthalene, naphthalene, 1,5-dimethyl-8ci 9ci
PubChem CID 11306
ChEBI CHEBI:48608
IUPAC Name 1,5-dimethylnaphthalene
SMILES CC1=CC=CC2=C(C)C=CC=C12

Specifications

Melting Point 81°C
Color White-Yellow
Quantity 1 g
Formula Weight 156.23
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Dimethylnaphthalene
EINECSNumber (4)-0080
TSCA No