missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dimethylnaphthalene, 99%

Catalog No. p-7023360
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA16819MC 100 mg
AAA16819ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA16819MC Supplier Thermo Scientific Chemicals Supplier No. A16819MC

CAS: 571-61-9 | C12H12 | 156.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 571-61-9
Molecular Formula C12H12
Molecular Weight (g/mol) 156.23
MDL Number MFCD00004038
InChI Key SDDBCEWUYXVGCQ-UHFFFAOYSA-N
Synonym naphthalene, 1,5-dimethyl, 1,5-dmn, 1,5-dimethyl-naphthalene, acmc-209lwa, naphthalene,5-dimethyl, 1,5-dimethyl naphthalene, 1,5-dimethylnaphthalene, naphthalene, 1,5-dimethyl-8ci 9ci
PubChem CID 11306
ChEBI CHEBI:48608
IUPAC Name 1,5-dimethylnaphthalene
SMILES CC1=CC=CC2=C(C)C=CC=C12

Specifications

Melting Point 78°C to 82°C
Boiling Point 265°C to 266°C
Odor Odorless
Quantity 100 mg
Beilstein 2039843
Formula Weight 156.23
Percent Purity 99%
Chemical Name or Material 1,5-Dimethylnaphthalene
EINECSNumber 209-338-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only