missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dichloropentane 95.0+%, TCI America™

Catalog No. D039025ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D039025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D039025ML Supplier TCI America Supplier No. D039025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 628-76-2
Molecular Formula C5H10Cl2
Molecular Weight (g/mol) 141.035
MDL Number MFCD00001017
InChI Key LBKDGROORAKTLC-UHFFFAOYSA-N
Synonym pentamethylene chloride, pentane, 1,5-dichloro, pentamethylene dichloride, amylene chloride, unii-6n4a5v83we, 1,5-dichlorobutane, pentane,5-dichloro, acmc-1ay6s, 1,5-bis chloranyl pentane, wln: g5g
PubChem CID 12353
IUPAC Name 1,5-dichloropentane
SMILES C(CCCl)CCCl

Specifications

Color Yellow
Boiling Point 182°C
Quantity 25 mL
UN Number 1152
Formula Weight 141.04
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,5-Dichloropentane
RTECSNumber SA0350000
TSCA Yes