missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dichloropentane, 98%

Catalog No. AAA1243322
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1243322 100 g
AAA1243336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1243322 Supplier Thermo Scientific Chemicals Supplier No. A1243322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 628-76-2 | C5H10Cl2 | 141.035 g/mol

1,5-Dichloropentane is used in the preparation of symmetrical sulfide, tetrydrothiopyran (thiane) by reaction with sodium sulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Dichloropentane is used in the preparation of symmetrical sulfide, tetrydrothiopyran (thiane) by reaction with sodium sulfide.

Solubility
Miscible with ethanol, ether and chloroform. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 628-76-2
Molecular Formula C5H10Cl2
Molecular Weight (g/mol) 141.035
MDL Number MFCD00001017
InChI Key LBKDGROORAKTLC-UHFFFAOYSA-N
Synonym pentamethylene chloride, pentane, 1,5-dichloro, pentamethylene dichloride, amylene chloride, unii-6n4a5v83we, 1,5-dichlorobutane, pentane,5-dichloro, acmc-1ay6s, 1,5-bis chloranyl pentane, wln: g5g
PubChem CID 12353
IUPAC Name 1,5-dichloropentane
SMILES C(CCCl)CCCl

Specifications

Melting Point -72°C
Density 1.103
Boiling Point 182°C
Flash Point 26°C (78°F)
Refractive Index 1.457
Linear Formula Cl(CH2)5Cl
Quantity 100 g
UN Number UN1152
Beilstein 1732340
Solubility Information Miscible with ethanol,ether and chloroform. Immiscible with water.
Formula Weight 141.04
Percent Purity 98%
Chemical Name or Material 1,5-Dichloropentane
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DICHLOROPENTANES
EINECSNumber 211-053-6
RTECSNumber SA0350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only