missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,5-Dichloro-1,1,3,3,5,5-hexamethyltrisiloxane 98.0+%, TCI America™
$202.15 - $604.51
Chemical Identifiers
| CAS | 3582-71-6 |
|---|---|
| Molecular Formula | C6H18Cl2O2Si3 |
| Molecular Weight (g/mol) | 277.363 |
| MDL Number | MFCD00045147 |
| InChI Key | GJIYNWRLGOMDEX-UHFFFAOYSA-N |
| Synonym | 1,5-dichlorohexamethyltrisiloxane, 1,5-dichloro-1,1,3,3,5,5-hexamethyltrisiloxane, trisiloxane, 1,5-dichloro-1,1,3,3,5,5-hexamethyl, poly dimethylsiloxane , chlorine terminated, 2,6-dichloro-2,4,4,6-tetramethyl-3,5-dioxa-2,4,6-trisilaheptane, acmc-20ap1k, 1 5-dichlorohexamethyltrisiloxane, 1,5-dichloro-hexamethyltrisiloxane, 1,5-dichlorohexamethyl trisiloxane, trisiloxane,1,5-dichloro,hexamethyl |
| PubChem CID | 77131 |
| IUPAC Name | bis[[chloro(dimethyl)silyl]oxy]-dimethylsilane |
| SMILES | C[Si](C)(O[Si](C)(C)Cl)O[Si](C)(C)Cl |
Chemical Identifiers
| 3582-71-6 | |
| 277.363 | |
| GJIYNWRLGOMDEX-UHFFFAOYSA-N | |
| 77131 | |
| C[Si](C)(O[Si](C)(C)Cl)O[Si](C)(C)Cl |
| C6H18Cl2O2Si3 | |
| MFCD00045147 | |
| 1,5-dichlorohexamethyltrisiloxane, 1,5-dichloro-1,1,3,3,5,5-hexamethyltrisiloxane, trisiloxane, 1,5-dichloro-1,1,3,3,5,5-hexamethyl, poly dimethylsiloxane , chlorine terminated, 2,6-dichloro-2,4,4,6-tetramethyl-3,5-dioxa-2,4,6-trisilaheptane, acmc-20ap1k, 1 5-dichlorohexamethyltrisiloxane, 1,5-dichloro-hexamethyltrisiloxane, 1,5-dichlorohexamethyl trisiloxane, trisiloxane,1,5-dichloro,hexamethyl | |
| bis[[chloro(dimethyl)silyl]oxy]-dimethylsilane |
Specifications
| 3582-71-6 | |
| 184°C | |
| MFCD00045147 | |
| 2987 | |
| GJIYNWRLGOMDEX-UHFFFAOYSA-N | |
| bis[[chloro(dimethyl)silyl]oxy]-dimethylsilane | |
| 77131 | |
| ≥98.0% (GC,T) | |
| 1,5-Dichloro-1,1,3,3,5,5-hexamethyltrisiloxane |
| Colorless | |
| C6H18Cl2O2Si3 | |
| 5 g | |
| 1,5-dichlorohexamethyltrisiloxane, 1,5-dichloro-1,1,3,3,5,5-hexamethyltrisiloxane, trisiloxane, 1,5-dichloro-1,1,3,3,5,5-hexamethyl, poly dimethylsiloxane , chlorine terminated, 2,6-dichloro-2,4,4,6-tetramethyl-3,5-dioxa-2,4,6-trisilaheptane, acmc-20ap1k, 1 5-dichlorohexamethyltrisiloxane, 1,5-dichloro-hexamethyltrisiloxane, 1,5-dichlorohexamethyl trisiloxane, trisiloxane,1,5-dichloro,hexamethyl | |
| C[Si](C)(O[Si](C)(C)Cl)O[Si](C)(C)Cl | |
| 277.363 | |
| 277.36 | |
| Liquid |
Safety and Handling
EINECSNumber : (7)-0472
TSCA : Yes