missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dichloro-1,1,3,3,5,5-hexamethyltrisiloxane 98.0+%, TCI America™

Catalog No. D28255G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28255G 5 g
D282525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28255G Supplier TCI America Supplier No. D28255G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3582-71-6
Molecular Formula C6H18Cl2O2Si3
Molecular Weight (g/mol) 277.363
MDL Number MFCD00045147
InChI Key GJIYNWRLGOMDEX-UHFFFAOYSA-N
Synonym 1,5-dichlorohexamethyltrisiloxane, 1,5-dichloro-1,1,3,3,5,5-hexamethyltrisiloxane, trisiloxane, 1,5-dichloro-1,1,3,3,5,5-hexamethyl, poly dimethylsiloxane , chlorine terminated, 2,6-dichloro-2,4,4,6-tetramethyl-3,5-dioxa-2,4,6-trisilaheptane, acmc-20ap1k, 1 5-dichlorohexamethyltrisiloxane, 1,5-dichloro-hexamethyltrisiloxane, 1,5-dichlorohexamethyl trisiloxane, trisiloxane,1,5-dichloro,hexamethyl
PubChem CID 77131
IUPAC Name bis[[chloro(dimethyl)silyl]oxy]-dimethylsilane
SMILES C[Si](C)(O[Si](C)(C)Cl)O[Si](C)(C)Cl

Specifications

Color Colorless
Boiling Point 184°C
Quantity 5 g
UN Number 2987
Formula Weight 277.36
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1,5-Dichloro-1,1,3,3,5,5-hexamethyltrisiloxane
EINECSNumber (7)-0472
TSCA Yes