missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dibromopentane, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 155990010

 View more versions of this product

Catalog No. AC155990010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

1, 5-Dibromopentane
111-24-0
100.0
224°C
Authentic
Glass bottle
1.5112 to 1.5132
MFCD00000268
01, 131
pentamethylene bromide, pentane, 1,5-dibromo, pentamethylene dibromide, unii-b8q228qys1, pentane-1,5-dibromide, 1,5-dibromo-pentane, 1,5-pentane dibromide, pentane,1,5-dibromo, br ch2 5br, ccris 8918
IBODDUNKEPPBKW-UHFFFAOYSA-N
1,5-dibromopentane
8100
97%
97%
96.0
1.6800g/mL
79°C
97%
C5H10Br2
Br(CH2)5Br
1L
1.68
Solubility in water: insoluble. Other solubilities: soluble in benzene,chloroform,slightly soluble in carbon tetrachloride
C(CCBr)CCBr
229.94
229.94

Chemical Identifiers

111-24-0
229.94
IBODDUNKEPPBKW-UHFFFAOYSA-N
8100
C(CCBr)CCBr
C5H10Br2
MFCD00000268
pentamethylene bromide, pentane, 1,5-dibromo, pentamethylene dibromide, unii-b8q228qys1, pentane-1,5-dibromide, 1,5-dibromo-pentane, 1,5-pentane dibromide, pentane,1,5-dibromo, br ch2 5br, ccris 8918
1,5-dibromopentane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-849-7

RTECSNumber : SA0320000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only