missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,5-Dibromonaphthalene 98.0+%, TCI America™
$305.70 - $764.27
Chemical Identifiers
| CAS | 7351-74-8 |
|---|---|
| Molecular Formula | C10H6Br2 |
| Molecular Weight (g/mol) | 285.966 |
| MDL Number | MFCD00089984 |
| InChI Key | CZYAFTZIQWCKOI-UHFFFAOYSA-N |
| PubChem CID | 637960 |
| IUPAC Name | 1,5-dibromonaphthalene |
| SMILES | C1=CC2=C(C=CC=C2Br)C(=C1)Br |
Chemical Identifiers
| 7351-74-8 | |
| 285.966 | |
| CZYAFTZIQWCKOI-UHFFFAOYSA-N | |
| 1,5-dibromonaphthalene |
| C10H6Br2 | |
| MFCD00089984 | |
| 637960 | |
| C1=CC2=C(C=CC=C2Br)C(=C1)Br |
Specifications
| 7351-74-8 | |
| Yellow | |
| C10H6Br2 | |
| 1 g | |
| C1=CC2=C(C=CC=C2Br)C(=C1)Br | |
| 285.966 | |
| 285.97 | |
| Crystalline Powder |
| 130°C | |
| 326°C | |
| MFCD00089984 | |
| CZYAFTZIQWCKOI-UHFFFAOYSA-N | |
| 1,5-dibromonaphthalene | |
| 637960 | |
| ≥98.0% (GC) | |
| 1,5-Dibromonaphthalene |
Safety and Handling
TSCA : No