missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dibromo-1,1,3,3,5,5-hexafluoropentane, 97%

Catalog No. AAB2140504
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2140504 2 g
AAB2140509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2140504 Supplier Thermo Scientific Chemicals Supplier No. B2140504
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 371-83-5 | C5H4Br2F6 | 337.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 371-83-5
Molecular Formula C5H4Br2F6
Molecular Weight (g/mol) 337.89
MDL Number MFCD00236657
InChI Key XYMBGGTVDWPIBA-UHFFFAOYSA-N
Synonym pentane,1,5-dibromo-1,1,3,3,5,5-hexafluoro, 1,5-dibromo-1,1,3,3,5,5-hexafluoropentane
PubChem CID 2736777
IUPAC Name 1,5-dibromo-1,1,3,3,5,5-hexafluoropentane
SMILES FC(F)(Br)CC(F)(F)CC(F)(F)Br

Specifications

Density 2.003
Refractive Index 1.4632
Linear Formula BrCF2CH2CF2CH2CF2Br
Quantity 2 g
Formula Weight 337.89
Percent Purity 97%
Chemical Name or Material 1,5-Dibromo-1,1,3,3,5,5-hexafluoropentane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only