missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diazabicyclo[4.3.0]non-5-ene, 98%

Catalog No. AAA1343714
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1343714 25 g
AAA1343722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1343714 Supplier Thermo Scientific Chemicals Supplier No. A1343714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3001-72-7 | C7H12N2 | 124.19 g/mol

1,5-Diazabicyclo[4.3.0]non-5-ene is employed for dehydrohalogenation reactions and base-catalyzed rearrangements in organic synthesis. It is an amidine base used in organic synthesis. Further, it is used as a resin curing agent and polyurethane catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Diazabicyclo[4.3.0]non-5-ene is employed for dehydrohalogenation reactions and base-catalyzed rearrangements in organic synthesis. It is an amidine base used in organic synthesis. Further, it is used as a resin curing agent and polyurethane catalyst.

Solubility
Miscible with water.

Notes
Air, light and moisture sensitive. Hygroscopic. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acid chlorides, acid anhydrides, oxidizing agents and chloroformates.

Chemical Identifiers

CAS 3001-72-7
Molecular Formula C7H12N2
Molecular Weight (g/mol) 124.19
MDL Number MFCD00005554
InChI Key SGUVLZREKBPKCE-UHFFFAOYSA-N
Synonym 1,5-diazabicyclo 4.3.0 non-5-ene, 2,3,4,6,7,8-hexahydropyrrolo 1,2-a pyrimidine, unii-978m4ol12q, 2h,3h,4h,6h,7h,8h-pyrrolo 1,2-a pyrimidine, pyrrolo 1,2-a pyrimidine, 2,3,4,6,7,8-hexahydro, 1,5-diazabicyclo 4,3,0 non-5-ene, 1,5-diazabicyclo 4.3.0-5-nonene, 1,5-diazabicyclo-4.3.0 non-5-ene
PubChem CID 76349
IUPAC Name 2,3,4,6,7,8-hexahydropyrrolo[1,2-a]pyrimidine
SMILES C1CN2CCCN=C2C1

Specifications

Density 1.042
Boiling Point 95°C to 98°C (7 mmHg)
Flash Point 94°C (201°F)
Refractive Index 1.52
Quantity 25 g
UN Number UN3267
Beilstein 2417
Sensitivity Air sensitive; Hygroscopic
Solubility Information Miscible with water.
Formula Weight 124.19
Percent Purity 98%
Chemical Name or Material 1,5-Diazabicyclo[4.3.0]non-5-ene
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
EINECSNumber 221-087-3
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only