missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diazabicyclo[4.3.0]-5-nonene 98.0+%, TCI America™

Catalog No. p-7126671
Change view
Click to view available options
Quantity:
10 mL
25 mL
250 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D131310ML 10 mL
D131325ML 25 mL
D1313250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D131310ML Supplier TCI America Supplier No. D131310ML
Only null left

Chemical Identifiers

CAS 3001-72-7
Molecular Formula C7H12N2
Molecular Weight (g/mol) 124.19
MDL Number MFCD00005554
InChI Key SGUVLZREKBPKCE-UHFFFAOYSA-N
Synonym 1,5-diazabicyclo 4.3.0 non-5-ene, 2,3,4,6,7,8-hexahydropyrrolo 1,2-a pyrimidine, unii-978m4ol12q, 2h,3h,4h,6h,7h,8h-pyrrolo 1,2-a pyrimidine, pyrrolo 1,2-a pyrimidine, 2,3,4,6,7,8-hexahydro, 1,5-diazabicyclo 4,3,0 non-5-ene, 1,5-diazabicyclo 4.3.0-5-nonene, 1,5-diazabicyclo-4.3.0 non-5-ene
PubChem CID 76349
IUPAC Name 2H,3H,4H,6H,7H,8H-pyrrolo[1,2-a]pyrimidine
SMILES C1CN2CCCN=C2C1

Specifications

Color Yellow
Boiling Point 108°C
Quantity 10 mL
UN Number 3267
Formula Weight 124.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,5-Diazabicyclo[4.3.0]-5-nonene
EINECSNumber (5)-6217
TSCA No