missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,5-Diaminopentane Dihydrochloride 98.0+%, TCI America™
$154.71 - $492.11
Chemical Identifiers
| CAS | 1476-39-7 |
|---|---|
| Molecular Formula | C5H16Cl2N2 |
| Molecular Weight (g/mol) | 175.097 |
| MDL Number | MFCD00012527 |
| InChI Key | FGNLEIGUMSBZQP-UHFFFAOYSA-N |
| Synonym | Cadaverine Hydrochloride, Pentamethylenediamine Hydrochloride, 1,5-Pentanediamine Dihydrochloride |
| PubChem CID | 80282 |
| ChEBI | CHEBI:64701 |
| IUPAC Name | pentane-1,5-diamine;dihydrochloride |
| SMILES | C(CCN)CCN.Cl.Cl |
Chemical Identifiers
| 1476-39-7 | |
| 175.097 | |
| FGNLEIGUMSBZQP-UHFFFAOYSA-N | |
| 80282 | |
| pentane-1,5-diamine;dihydrochloride |
| C5H16Cl2N2 | |
| MFCD00012527 | |
| Cadaverine Hydrochloride, Pentamethylenediamine Hydrochloride, 1,5-Pentanediamine Dihydrochloride | |
| CHEBI:64701 | |
| C(CCN)CCN.Cl.Cl |
Specifications
| 1476-39-7 | |
| White-Yellow | |
| MFCD00012527 | |
| Cadaverine Hydrochloride, Pentamethylenediamine Hydrochloride, 1,5-Pentanediamine Dihydrochloride | |
| C(CCN)CCN.Cl.Cl | |
| 175.097 | |
| CHEBI:64701 | |
| ≥98.0% (N) | |
| 1,5-Diaminopentane Dihydrochloride |
| 230°C | |
| C5H16Cl2N2 | |
| 5 g | |
| FGNLEIGUMSBZQP-UHFFFAOYSA-N | |
| pentane-1,5-diamine;dihydrochloride | |
| 80282 | |
| 175.10 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes