missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diaminopentane Dihydrochloride 98.0+%, TCI America™

Catalog No. D00995G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D00995G 5 g
D009925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D00995G Supplier TCI America Supplier No. D00995G
Only null left

Chemical Identifiers

CAS 1476-39-7
Molecular Formula C5H16Cl2N2
Molecular Weight (g/mol) 175.097
MDL Number MFCD00012527
InChI Key FGNLEIGUMSBZQP-UHFFFAOYSA-N
Synonym Cadaverine Hydrochloride, Pentamethylenediamine Hydrochloride, 1,5-Pentanediamine Dihydrochloride
PubChem CID 80282
ChEBI CHEBI:64701
IUPAC Name pentane-1,5-diamine;dihydrochloride
SMILES C(CCN)CCN.Cl.Cl

Specifications

Melting Point 230°C
Color White-Yellow
Quantity 5 g
Formula Weight 175.10
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Diaminopentane Dihydrochloride