missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diaminonaphthalene 98.0+%, TCI America™

Catalog No. D010125G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D010125G 25 g
D0101100G 100 g
D0101500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D010125G Supplier TCI America Supplier No. D010125G

Chemical Identifiers

CAS 2243-62-1
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.204
MDL Number MFCD00004029
InChI Key KQSABULTKYLFEV-UHFFFAOYSA-N
Synonym 1,5-diaminonaphthalene, 1,5-naphthalenediamine, 1,5-naphthylenediamine, 1,5-napthalenediamine, unii-13pd3j52lk, ccris 422, 1,5-diamino naphthalene, 1,5-diaminonaphtalene, dsstox_cid_916, acmc-1crp7
PubChem CID 16720
ChEBI CHEBI:53003
IUPAC Name naphthalene-1,5-diamine
SMILES C1=CC2=C(C=CC=C2N)C(=C1)N

Specifications

Melting Point 187°C
Boiling Point 210°C
Quantity 25 g
UN Number 3077
Formula Weight 158.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Diaminonaphthalene
EINECSNumber (4)-0323
RTECSNumber QJ3400000
TSCA No