missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diaminonaphthalene, 97%

Catalog No. AC112291000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC112291000 100 g Glass bottle
AC112290250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC112291000 Supplier Thermo Scientific Chemicals Supplier No. 112291000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2243-62-1 | C10H10N2 | 158.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2243-62-1
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.2
MDL Number MFCD00004029
InChI Key KQSABULTKYLFEV-UHFFFAOYSA-N
Synonym 1,5-diaminonaphthalene, 1,5-naphthalenediamine, 1,5-naphthylenediamine, 1,5-napthalenediamine, unii-13pd3j52lk, ccris 422, 1,5-diamino naphthalene, 1,5-diaminonaphtalene, dsstox_cid_916, acmc-1crp7
PubChem CID 16720
ChEBI CHEBI:53003
IUPAC Name naphthalene-1,5-diamine
SMILES C1=CC2=C(C=CC=C2N)C(=C1)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 187°C to 190°C
Color Brown to Gray
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C10H6(NH2)2
Quantity 100 g
Beilstein 13, 203
Solubility Information Solubility in water: soluble in hot water. Other solubilities: sparingly soluble in cold water,soluble in alcohol
Formula Weight 158.2
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1, 5-Diaminonaphthalene, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Suspected of causing cancer.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 218-817-8
RTECSNumber QJ3400000
TSCA TSCA

RUO – Research Use Only