missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diaminonaphthalene, 97%

Catalog No. AAB2426222
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2426222 100 g
AAB2426214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2426222 Supplier Thermo Scientific Chemicals Supplier No. B2426222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2243-62-1 | C10H10N2 | 158.204 g/mol

t is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. The reducing properties of 1,5-diaminonaphthalene (1,5-DAN) as a MALDI matrix ( matrix-assisted laser desorption ionization) to amino acid sequencing and disulfide bond mapping of human urotensin II possessing one disulfide bond, and human guanylin possessing two disulfide bonds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
t is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. The reducing properties of 1,5-diaminonaphthalene (1,5-DAN) as a MALDI matrix ( matrix-assisted laser desorption ionization) to amino acid sequencing and disulfide bond mapping of human urotensin II possessing one disulfide bond, and human guanylin possessing two disulfide bonds.

Solubility
Soluble in water. (0.3 g/L)

Notes
Store at room temperature. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2243-62-1
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.204
MDL Number MFCD00004029
InChI Key KQSABULTKYLFEV-UHFFFAOYSA-N
Synonym 1,5-diaminonaphthalene, 1,5-naphthalenediamine, 1,5-naphthylenediamine, 1,5-napthalenediamine, unii-13pd3j52lk, ccris 422, 1,5-diamino naphthalene, 1,5-diaminonaphtalene, dsstox_cid_916, acmc-1crp7
PubChem CID 16720
ChEBI CHEBI:53003
IUPAC Name naphthalene-1,5-diamine
SMILES C1=CC2=C(C=CC=C2N)C(=C1)N

Specifications

Melting Point 189°C to 192°C
Boiling Point 200°C to 210°C (5 mmHg)
Quantity 100 g
UN Number UN3077
Beilstein 907947
Solubility Information Soluble in water. (0.3g/L)
Formula Weight 158.2
Percent Purity 97%
Chemical Name or Material 1,5-Diaminonaphthalene
Hazard Category H302-H312-H351
Hazard Statement GHS H Statement
H351
Suspected of causing cancer.
Precautionary Statement P201-P202-P264c-P270-P280-P301+P310-P302+P352-P308+P313-P312-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 218-817-8
RTECSNumber QJ3400000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only