missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diaminoanthraquinone 92.0+%, TCI America™

Catalog No. D12685G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D12685G Supplier TCI America Supplier No. D12685G

Chemical Identifiers

CAS 129-44-2
Molecular Formula C14H10N2O2
Molecular Weight (g/mol) 238.246
MDL Number MFCD00001226
InChI Key VWBVCOPVKXNMMZ-UHFFFAOYSA-N
Synonym 1,5-diaminoanthraquinone, smoke red f, 1,5-diaminoanthrachinon, c.i. disperse red ii, 1,5-anthraquinonyldiamine, 9,10-anthracenedione, 1,5-diamino, 1,5-diamino-9,10-anthraquinone, 1,5-daa, 1,5-daa russian, unii-3zxx3hk358
PubChem CID 8513
IUPAC Name 1,5-diaminoanthracene-9,10-dione
SMILES C1=CC2=C(C(=C1)N)C(=O)C3=C(C2=O)C(=CC=C3)N

Specifications

Quantity 5 g
Formula Weight 238.25
Percent Purity ≥92.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Diaminoanthraquinone
EINECSNumber (4)-0710
RTECSNumber CB6400000
TSCA Yes