Learn More
1,5-Diaminoanthraquinone, 90+%
CAS: 129-44-2 | C14H10N2O2 | 238.246 g/mol
Supplier: Thermo Scientific Chemicals A1884022
Description
1,5-Diaminoanthraquinone (DAAQ) may be used in the preparation of vertical nanowire arrays. It may be used in the synthesis of poly(1,5-diaminoanthraquinone) (PDAA) particles, via chemically oxidative polymerization of DAAQ.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,5-Diaminoanthraquinone (DAAQ) may be used in the preparation of vertical nanowire arrays. It may be used in the synthesis of poly(1,5-diaminoanthraquinone) (PDAA) particles, via chemically oxidative polymerization of DAAQ.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
| 1,5-Diaminoanthraquinone | |
| ∼320°C (decomposition) | |
| C14H10N2O2 | |
| 100 g | |
| Insoluble in water. | |
| C1=CC2=C(C(=C1)N)C(=O)C3=C(C2=O)C(=CC=C3)N | |
| 238.246 | |
| 238.25 |
| 129-44-2 | |
| Odorless | |
| MFCD00001226 | |
| 1,5-diaminoanthraquinone, smoke red f, 1,5-diaminoanthrachinon, c.i. disperse red ii, 1,5-anthraquinonyldiamine, 9,10-anthracenedione, 1,5-diamino, 1,5-diamino-9,10-anthraquinone, 1,5-daa, 1,5-daa russian, unii-3zxx3hk358 | |
| VWBVCOPVKXNMMZ-UHFFFAOYSA-N | |
| 1,5-diaminoanthracene-9,10-dione | |
| 8513 | |
| 97% |
Chemical Identifiers
| 129-44-2 | |
| 238.246 | |
| VWBVCOPVKXNMMZ-UHFFFAOYSA-N | |
| 8513 | |
| C1=CC2=C(C(=C1)N)C(=O)C3=C(C2=O)C(=CC=C3)N |
| C14H10N2O2 | |
| MFCD00001226 | |
| 1,5-diaminoanthraquinone, smoke red f, 1,5-diaminoanthrachinon, c.i. disperse red ii, 1,5-anthraquinonyldiamine, 9,10-anthracenedione, 1,5-diamino, 1,5-diamino-9,10-anthraquinone, 1,5-daa, 1,5-daa russian, unii-3zxx3hk358 | |
| 1,5-diaminoanthracene-9,10-dione |
Safety and Handling
EINECSNumber : 204-947-2
RTECSNumber : CB6400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only