missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diaminoanthraquinone, 90+%

Catalog No. AAA1884014
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1884014 25 g
AAA1884022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1884014 Supplier Thermo Scientific Chemicals Supplier No. A1884014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 129-44-2 | C14H10N2O2 | 238.246 g/mol

1,5-Diaminoanthraquinone (DAAQ) may be used in the preparation of vertical nanowire arrays. It may be used in the synthesis of poly(1,5-diaminoanthraquinone) (PDAA) particles, via chemically oxidative polymerization of DAAQ.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Diaminoanthraquinone (DAAQ) may be used in the preparation of vertical nanowire arrays. It may be used in the synthesis of poly(1,5-diaminoanthraquinone) (PDAA) particles, via chemically oxidative polymerization of DAAQ.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 129-44-2
Molecular Formula C14H10N2O2
Molecular Weight (g/mol) 238.246
MDL Number MFCD00001226
InChI Key VWBVCOPVKXNMMZ-UHFFFAOYSA-N
Synonym 1,5-diaminoanthraquinone, smoke red f, 1,5-diaminoanthrachinon, c.i. disperse red ii, 1,5-anthraquinonyldiamine, 9,10-anthracenedione, 1,5-diamino, 1,5-diamino-9,10-anthraquinone, 1,5-daa, 1,5-daa russian, unii-3zxx3hk358
PubChem CID 8513
IUPAC Name 1,5-diaminoanthracene-9,10-dione
SMILES C1=CC2=C(C(=C1)N)C(=O)C3=C(C2=O)C(=CC=C3)N

Specifications

Melting Point ∼320°C (decomposition)
Odor Odorless
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 238.25
Percent Purity 97%
Chemical Name or Material 1,5-Diaminoanthraquinone
EINECSNumber 204-947-2
RTECSNumber CB6400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only