missing translation for 'onlineSavingsMsg'
Learn More

1,5-Bis(methylamino)-3-oxapentane, 98%, Thermo Scientific™

Chemical Identifiers

CAS 2620-27-1
Molecular Formula C6H16N2O
Molecular Weight (g/mol) 132.21
InChI Key VXPJBVRYAHYMNY-UHFFFAOYSA-N
Synonym 2,2'-oxybis n-methylethanamine, ethanamine,2,2'-oxybis n-methyl, ethanamine, 2,2'-oxybis n-methyl, 1,5-bis methylamino-3-oxapentane, methyl 2-2-methylamino ethoxy ethyl amine, bmaop, n-methyl-2-methylaminoethyloxy ethylamine, n-methyl-2-2-methylamino ethoxy ethanamine, n-methyl-n-2-2-methylamino ethoxy ethyl amine
PubChem CID 2724770
IUPAC Name N-methyl-2-[2-(methylamino)ethoxy]ethanamine
SMILES CNCCOCCNC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC275860050
SDS View Documents
Thermo Scientific Chemicals
275860050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2620-27-1
132.21
2,2'-oxybis n-methylethanamine, ethanamine,2,2'-oxybis n-methyl, ethanamine, 2,2'-oxybis n-methyl, 1,5-bis methylamino-3-oxapentane, methyl 2-2-methylamino ethoxy ethyl amine, bmaop, n-methyl-2-methylaminoethyloxy ethylamine, n-methyl-2-2-methylamino ethoxy ethanamine, n-methyl-n-2-2-methylamino ethoxy ethyl amine
N-methyl-2-[2-(methylamino)ethoxy]ethanamine
C6H16N2O
VXPJBVRYAHYMNY-UHFFFAOYSA-N
2724770
CNCCOCCNC

Specifications

2620-27-1
76.0°C to 78.0°C (20.0mmHg)
98%
C6H16N2O
1.4370 to 1.4390
0.872
Solubility in water: soluble. Other solubilities: soluble in methanol,chloroform
CNCCOCCNC
132.21
132.21
1, 5-Bis(methylamino)-3-oxapentane
0.8720g/mL
Authentic
Glass bottle
CH3NHCH2CH2OCH2CH2NHCH3
5g
2,2'-oxybis n-methylethanamine, ethanamine,2,2'-oxybis n-methyl, ethanamine, 2,2'-oxybis n-methyl, 1,5-bis methylamino-3-oxapentane, methyl 2-2-methylamino ethoxy ethyl amine, bmaop, n-methyl-2-methylaminoethyloxy ethylamine, n-methyl-2-2-methylamino ethoxy ethanamine, n-methyl-n-2-2-methylamino ethoxy ethyl amine
VXPJBVRYAHYMNY-UHFFFAOYSA-N
N-methyl-2-[2-(methylamino)ethoxy]ethanamine
2724770
98%
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only