missing translation for 'onlineSavingsMsg'
Learn More

1,5-Bis(diphenylphosphino)pentane, 97%

Catalog No. AAB2105403
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2105403 1 g
AAB2105406 5 g
AAB2105414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2105403 Supplier Thermo Scientific Chemicals Supplier No. B2105403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 27721-02-4 | C29H30P2 | 440.51 g/mol

1,5-Bis(diphenylphosphino)pentane is used as a ligand for the cross coupling reactions. It is used in the manufacturing of glass bottle PTFE bottle, aluminum foil bag and in cardboard drum. combination of a 1,5-bis(diphenylphosphino)pentane ligand and the addition of N,N,N?,N?-tetramethylethylenediamine (TMEDA) as a co-catalyst are the key factors in obtaining the corresponding aryl nitriles with improved catalyst productivities and selectivities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Bis(diphenylphosphino)pentane is used as a ligand for the cross coupling reactions. It is used in the manufacturing of glass bottle PTFE bottle, aluminum foil bag and in cardboard drum. combination of a 1,5-bis(diphenylphosphino)pentane ligand and the addition of N,N,N′,N′-tetramethylethylenediamine (TMEDA) as a co-catalyst are the key factors in obtaining the corresponding aryl nitriles with improved catalyst productivities and selectivities.

Solubility
Soluble in N,N-dimethyl formamide.

Notes
Store at room temperature. Keep away from oxidizing agents.

Chemical Identifiers

CAS 27721-02-4
Molecular Formula C29H30P2
Molecular Weight (g/mol) 440.51
MDL Number MFCD00003052
InChI Key MZFPAWGWFDGCHP-UHFFFAOYSA-N
Synonym 1,5-bis diphenylphosphino pentane, dpppe, dppent, 5-diphenylphosphanylpentyl diphenyl phosphane, pentamethylenebis diphenylphosphine, 1,5-pentanediylbis diphenylphosphine, phosphine, 1,5-pentanediylbis diphenyl, 1-5-diphenylphosphino pentyl phenyl phosphino benzene
PubChem CID 2733414
SMILES C(CCP(C1=CC=CC=C1)C1=CC=CC=C1)CCP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 46°C to 48°C
Quantity 1 g
Solubility Information Soluble in N,N-dimethyl formamide.
IUPAC Name 5-diphenylphosphanylpentyl(diphenyl)phosphane
Formula Weight 440.5
Percent Purity 97%
Chemical Name or Material 1,5-Bis(diphenylphosphino)pentane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only