missing translation for 'onlineSavingsMsg'
Learn More

1,5,7-Triazabicyclo[4.4.0]dec-5-ene 98.0+%, TCI America™

Catalog No. p-7137049
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19825G 5 g
T198225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T19825G Supplier TCI America Supplier No. T19825G

Chemical Identifiers

CAS 5807-14-7
Molecular Formula C7H13N3
Molecular Weight (g/mol) 139.202
MDL Number MFCD00043003
InChI Key FVKFHMNJTHKMRX-UHFFFAOYSA-N
Synonym 1,3,4,6,7,8-Hexahydro-2H-pyrimido[1,2-a]pyrimidine, TBD
PubChem CID 79873
IUPAC Name 3,4,6,7,8,9-hexahydro-2H-pyrimido[1,2-a]pyrimidine
SMILES C1CNC2=NCCCN2C1

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 5 g
UN Number 3263
Formula Weight 139.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,5,7-Triazabicyclo[4.4.0]dec-5-ene