missing translation for 'onlineSavingsMsg'
Learn More

1,5,5-Trimethylhydantoin, 98%

Catalog No. p-7027439
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2418806 5 g
AAB2418814 25 g
AAB2418822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2418806 Supplier Thermo Scientific Chemicals Supplier No. B2418806

CAS: 6851-81-6 | C6H10N2O2 | 142.158 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6851-81-6
Molecular Formula C6H10N2O2
Molecular Weight (g/mol) 142.158
MDL Number MFCD00040439
InChI Key ZNYIPTYJBRGSSL-UHFFFAOYSA-N
Synonym 1,5,5-trimethylhydantoin, 3,4,4-trimethyl-2,5-dioxoimidazolidine, 1,5,5-trimethyl-imidazolidine-2,4-dione, 1,5,5-trimethyl-1,3-diazolidine-2,4-dione, hydantoin, 1,5,5-trimethyl, 2,4-imidazolidinedione, 1,5,5-trimethyl, 1,5,5 trimethylhydantoin, acmc-209o3u, 1,5,5-trimethyl-hydantoin, 1,5,5-trimethyl hydantoin
PubChem CID 81295
IUPAC Name 1,5,5-trimethylimidazolidine-2,4-dione
SMILES CC1(C(=O)NC(=O)N1C)C

Specifications

Melting Point 161°C to 164°C
Quantity 5 g
Formula Weight 142.16
Percent Purity 98%
Chemical Name or Material 1,5,5-Trimethylhydantoin
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 229-945-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only