missing translation for 'onlineSavingsMsg'
Learn More

1-[4-(Trifluoromethyl)pyrimidin-2-yl]-1,4-diazepane, 95%, Thermo Scientific™

Catalog No. p-2695413
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AW00488DA 1g
AW00488DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AW00488DA Supplier Thermo Scientific Chemicals Supplier No. AW00488DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 238403-48-0
Molecular Formula C10H13F3N4
Molecular Weight (g/mol) 246.24
MDL Number MFCD00728997
InChI Key XVAZHAVSZFQOOQ-UHFFFAOYSA-N
Synonym 1-4-trifluoromethyl pyrimidin-2-yl-1,4-diazepane, 1-4-trifluoromethyl-2-pyrimidinyl-1,4-diazepane, 1-4-trifluoromethyl pyrimid-2-yl homopiperazine, 1-4-trifluoromethyl pyrimidin-2-yl homopiperazine, 1h-1,4-diazepine,hexahydro-1-4-trifluoromethyl-2-pyrimidinyl, maybridge1_000269, 1-4-trifluoromethyl pyrimidin-2-yl-1,4-diazaperhydroepine
PubChem CID 2782067
IUPAC Name 1-[4-(trifluoromethyl)pyrimidin-2-yl]-1,4-diazepane
SMILES FC(F)(F)C1=NC(=NC=C1)N1CCCNCC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1g
Formula Weight 246.23
Percent Purity 95%
Chemical Name or Material 1-[4-(Trifluoromethyl)pyrimidin-2-yl]-1, 4-diazepane
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.