Learn More
1-[4-(Trifluoromethyl)-2-pyrimidinyl]-4-piperidinecarboxylic acid, ≥97%, Thermo Scientific™
Chemical Identifiers
| CAS | 465514-39-0 |
|---|---|
| Molecular Formula | C11H12F3N3O2 |
| Molecular Weight (g/mol) | 275.23 |
| MDL Number | MFCD02091615 |
| InChI Key | QYAIISDWPUEPHG-UHFFFAOYSA-N |
| Synonym | 1-4-trifluoromethyl-2-pyrimidinyl-4-piperidinecarboxylic acid, 1-4-trifluoromethyl pyrimidin-2-yl piperidine-4-carboxylic acid, 1-4-trifluoromethyl-2-pyrimidinyl-4-piperidine carboxylic acid, 1-4-trifluoromethyl pyrimid-2-yl piperidine-4-carboxylic acid, 1-4-trifluoromethyl-pyrimidin-2-yl-4-piperidinecarboxylic acid |
| PubChem CID | 2777798 |
| IUPAC Name | 1-[4-(trifluoromethyl)pyrimidin-2-yl]piperidine-4-carboxylic acid |
| SMILES | OC(=O)C1CCN(CC1)C1=NC=CC(=N1)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AW01250DA
|
Thermo Scientific Chemicals
AW01250DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AW01250EA
|
Thermo Scientific Chemicals
AW01250EA |
10g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 465514-39-0 | |
| 275.23 | |
| QYAIISDWPUEPHG-UHFFFAOYSA-N | |
| 2777798 | |
| OC(=O)C1CCN(CC1)C1=NC=CC(=N1)C(F)(F)F |
| C11H12F3N3O2 | |
| MFCD02091615 | |
| 1-4-trifluoromethyl-2-pyrimidinyl-4-piperidinecarboxylic acid, 1-4-trifluoromethyl pyrimidin-2-yl piperidine-4-carboxylic acid, 1-4-trifluoromethyl-2-pyrimidinyl-4-piperidine carboxylic acid, 1-4-trifluoromethyl pyrimid-2-yl piperidine-4-carboxylic acid, 1-4-trifluoromethyl-pyrimidin-2-yl-4-piperidinecarboxylic acid | |
| 1-[4-(trifluoromethyl)pyrimidin-2-yl]piperidine-4-carboxylic acid |
Specifications
| 465514-39-0 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD02091615 | |
| 1-4-trifluoromethyl-2-pyrimidinyl-4-piperidinecarboxylic acid, 1-4-trifluoromethyl pyrimidin-2-yl piperidine-4-carboxylic acid, 1-4-trifluoromethyl-2-pyrimidinyl-4-piperidine carboxylic acid, 1-4-trifluoromethyl pyrimid-2-yl piperidine-4-carboxylic acid, 1-4-trifluoromethyl-pyrimidin-2-yl-4-piperidinecarboxylic acid | |
| OC(=O)C1CCN(CC1)C1=NC=CC(=N1)C(F)(F)F | |
| 275.23 | |
| 275.23 | |
| 1-[4-(Trifluoromethyl)-2-pyrimidinyl]-4-piperidinecarboxylic acid |
| 96.5 | |
| 97% | |
| C11H12F3N3O2 | |
| 1g | |
| QYAIISDWPUEPHG-UHFFFAOYSA-N | |
| 1-[4-(trifluoromethyl)pyrimidin-2-yl]piperidine-4-carboxylic acid | |
| 2777798 | |
| 97% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.