missing translation for 'onlineSavingsMsg'
Learn More

1-(4-tert-Butylphenyl)-3-(4-methoxyphenyl)-1,3-propanedione 98.0+%, TCI America™

Catalog No. p-7122912
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B338225G 25 g
B3382100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B338225G Supplier TCI America Supplier No. B338225G
Only null left

Chemical Identifiers

CAS 70356-09-1
Molecular Formula C20H22O3
Molecular Weight (g/mol) 310.39
MDL Number MFCD00210252
InChI Key XNEFYCZVKIDDMS-UHFFFAOYSA-N
PubChem CID 51040
IUPAC Name 1-(4-tert-butylphenyl)-3-(4-methoxyphenyl)propane-1,3-dione
SMILES COC1=CC=C(C=C1)C(=O)CC(=O)C1=CC=C(C=C1)C(C)(C)C

Specifications

Melting Point 82°C
Color White-Yellow
Quantity 25 g
Formula Weight 310.39
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-tert-Butylphenyl)-3-(4-methoxyphenyl)-1,3-propanedione