missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Propylphenyl)-2,5-di(2-thienyl)-1H-pyrrole 98.0+%, TCI America™

Catalog No. P2210200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2210200MG 200 mg
P22101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2210200MG Supplier TCI America Supplier No. P2210200MG
Only null left

Chemical Identifiers

CAS 499793-84-9
Molecular Formula C21H19NS2
Molecular Weight (g/mol) 349.51
MDL Number MFCD30721945
InChI Key JFTYJFYWQQLLFG-UHFFFAOYSA-N
PubChem CID 132274863
IUPAC Name 1-(4-propylphenyl)-2,5-bis(thiophen-2-yl)-1H-pyrrole
SMILES CCCC1=CC=C(C=C1)N1C(=CC=C1C1=CC=CS1)C1=CC=CS1

Specifications

Melting Point 147°C
Color Yellow
Quantity 200 mg
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Propylphenyl)-2,5-di(2-thienyl)-1H-pyrrole