missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Phenylenediboronic Acid (contains varying amounts of Anhydride), TCI America™
Specifications
Specifications
| Quantity | 5 g |
| Formula Weight | 165.75 |
| Physical Form | Crystalline Powder |
Safety and Handling
| TSCA | No |