missing translation for 'onlineSavingsMsg'
Learn More

1,4-Phenylenediamine Dihydrochloride 99.0+%, TCI America™

Catalog No. P015225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P015225G 25 g
P0152500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P015225G Supplier TCI America Supplier No. P015225G
Only null left

Chemical Identifiers

CAS 624-18-0
Molecular Formula C6H10Cl2N2
Molecular Weight (g/mol) 181.06
MDL Number MFCD00013002
InChI Key IYXMNTLBLQNMLM-UHFFFAOYSA-N
Synonym benzene-1,4-diamine dihydrochloride, p-phenylenediamine dihydrochloride, 1,4-diaminobenzene dihydrochloride, 1,4-phenylenediamine dihydrochloride, fourrine ds, 1,4-benzenediamine, dihydrochloride, pelagol cd, pelagol grey cd, durafur black rc, fourrine 64
PubChem CID 12205
IUPAC Name benzene-1,4-diamine;dihydrochloride
SMILES C1=CC(=CC=C1N)N.Cl.Cl

Specifications

Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 181.06
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Phenylenediamine Dihydrochloride
EINECSNumber (3)-0185&(5)-3029
RTECSNumber ST0350000
TSCA Yes