missing translation for 'onlineSavingsMsg'
Learn More

1,4-Phenylenediacetic Acid 98.0+%, TCI America™

Catalog No. P14335G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14335G 5 g
P143325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P14335G Supplier TCI America Supplier No. P14335G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7325-46-4
Molecular Formula C10H8O4
Molecular Weight (g/mol) 192.17
MDL Number MFCD00004354
InChI Key SLWIPPZWFZGHEU-UHFFFAOYSA-L
Synonym 1,4-phenylenediacetic acid, p-phenylenediacetic acid, 2,2'-1,4-phenylene diacetic acid, 1,4-benzenediacetic acid, 2-4-carboxymethyl phenyl acetic acid, 4-carboxymethyl phenyl acetic acid, p-benzenediacetic acid, acmc-209oq4, 1.4-phenylenediacetic acid, ksc378o7r
PubChem CID 81760
IUPAC Name 2-[4-(carboxylatomethyl)phenyl]acetate
SMILES [O-]C(=O)CC1=CC=C(CC([O-])=O)C=C1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 194.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Phenylenediacetic Acid