missing translation for 'onlineSavingsMsg'
Learn More

1,4-Phenylenediacetic acid, 97%

Catalog No. p-7023016
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1603603 1 g
AAA1603606 5 g
AAA1603614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1603603 Supplier Thermo Scientific Chemicals Supplier No. A1603603

CAS: 7325-46-4 | C10H8O4 | 192.17 g/mol

1,4-Phenylenediacetic acid is an arenediacetic acid used in the preparation of flurorescent whitening agents. It has fluorescent properties that allow for use in fluorescent probes. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Phenylenediacetic acid is an arenediacetic acid used in the preparation of flurorescent whitening agents. It has fluorescent properties that allow for use in fluorescent probes. It is also used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 7325-46-4
Molecular Formula C10H8O4
Molecular Weight (g/mol) 192.17
MDL Number MFCD00004354
InChI Key SLWIPPZWFZGHEU-UHFFFAOYSA-L
Synonym 1,4-phenylenediacetic acid, p-phenylenediacetic acid, 2,2'-1,4-phenylene diacetic acid, 1,4-benzenediacetic acid, 2-4-carboxymethyl phenyl acetic acid, 4-carboxymethyl phenyl acetic acid, p-benzenediacetic acid, acmc-209oq4, 1.4-phenylenediacetic acid, ksc378o7r
PubChem CID 81760
IUPAC Name 2-[4-(carboxymethyl)phenyl]acetic acid
SMILES [O-]C(=O)CC1=CC=C(CC([O-])=O)C=C1

Specifications

Melting Point 252°C to 256°C
Odor Odorless
Quantity 1 g
Beilstein 1960342
Solubility Information Slightly soluble in water.
Formula Weight 194.19
Percent Purity 97%
Chemical Name or Material 1,4-Phenylenediacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 230-801-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only