missing translation for 'onlineSavingsMsg'
Learn More

1,4-Pentadiene 98.0+%, TCI America™

Catalog No. P003825ML
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P003825ML 25 mL
P003810ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P003825ML Supplier TCI America Supplier No. P003825ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 591-93-5
Molecular Formula C5H8
Molecular Weight (g/mol) 68.119
MDL Number MFCD00008665
InChI Key QYZLKGVUSQXAMU-UHFFFAOYSA-N
Synonym 1,4-pentadiene, unii-a5if861oq5, pentadiene-1,4, allylethylene, penta-1,4-dien-1-yl, penta-1,4-dien-3-yl, ch2=chch2ch=ch2, acmc-1at58
PubChem CID 11587
IUPAC Name penta-1,4-diene
SMILES C=CCC=C

Specifications

Color Colorless
Boiling Point 26°C
Quantity 25 mL
UN Number 3295
Formula Weight 68.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Pentadiene
EINECSNumber (2)-0020
TSCA Yes
Recommended Storage Freezer