missing translation for 'onlineSavingsMsg'
Learn More

1,4-Pentadien-3-ol, 97%, stabilized

Catalog No. p-4089544
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC426240050 5 g Glass bottle
AC426240250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC426240050 Supplier Thermo Scientific Chemicals Supplier No. 426240050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 922-65-6 | C5H8O | 84.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 922-65-6
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00008633
InChI Key ICMWSAALRSINTC-UHFFFAOYSA-N
Synonym 1,4-pentadien-3-ol, ccris 8963, divinylcarbinol, divinyl carbinol, acmc-20ap1x, 1,4-pentadien-3-ol, contains hydroquinone as stabilizer
PubChem CID 70204
IUPAC Name penta-1,4-dien-3-ol
SMILES OC(C=C)C=C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.8650g/mL
Boiling Point 115.0°C
Flash Point 30°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4440 to 1.4460 (20°C, 589nm)
Linear Formula (CH2CH)2CHOH
Quantity 5 g
Specific Gravity 0.865
Stabilizer approximately 0.4% hydroquinone
Formula Weight 84.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 4-Pentadien-3-ol, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
EINECSNumber 213-080-9

RUO – Research Use Only