missing translation for 'onlineSavingsMsg'
Learn More

1,4-Oxazepan-5-one, 97%

Catalog No. p-7026417
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2240603 1 g
AAB2240606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2240603 Supplier Thermo Scientific Chemicals Supplier No. B2240603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10341-26-1 | C5H9NO2 | 115.13 g/mol

Used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a pharmaceutical intermediate.

Notes
Hygroscopic. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 10341-26-1
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.13
MDL Number MFCD02683091
InChI Key OXDIYMHNQAWSCL-UHFFFAOYSA-N
Synonym tetrahydro-1,4-oxazepin-5 2h-one, 1,4-oxazepin-5 2h-one, tetrahydro, 4-oxacaprolactam, acmc-1bsly, tetrahydro-1,4-oxazepin-5 4h-one, tetrahydro-2h-1,4-oxazepin-5-one
PubChem CID 7023020
IUPAC Name 1,4-oxazepan-5-one
SMILES O=C1CCOCCN1

Specifications

Melting Point 80°C to 82°C
Quantity 1 g
Sensitivity Hygroscopic
Formula Weight 115.13
Percent Purity 97%
Chemical Name or Material 1,4-Oxazepan-5-one
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No

RUO – Research Use Only