missing translation for 'onlineSavingsMsg'
Learn More

1,4-Oxathiane, 98+%

Catalog No. p-7023964
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1805206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1805206 Supplier Thermo Scientific Chemicals Supplier No. A1805206

CAS: 15980-15-1 | C4H8OS | 104.167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15980-15-1
Molecular Formula C4H8OS
Molecular Weight (g/mol) 104.167
MDL Number MFCD00006573
InChI Key JBYHSSAVUBIJMK-UHFFFAOYSA-N
Synonym 1,4-thioxane, p-thioxane, 1-oxa-4-thiacyclohexane, ccris 1746, 1,4-oxathiin, 2,3,5,6-tetrahydro, thioather, 1 pound not4-thioxane, 1-thia-4-oxacyclohexane, acmc-1bt7b, wln: t6o dstj
PubChem CID 27596
IUPAC Name 1,4-oxathiane
SMILES C1CSCCO1

Specifications

Density 1.114
Boiling Point 147°C to 148°C
Flash Point 42°C (107°F)
Odor Unpleasant
Refractive Index 1.507
Quantity 5 g
UN Number UN1993
Beilstein 102552
Formula Weight 104.17
Percent Purity ≥98%
Chemical Name or Material 1,4-Oxathiane
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226-H319
Flammable liquid and vapour.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 240-117-6
RTECSNumber RP4200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only