missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Nitrophenyl)pyrrole 98.0+%, TCI America™

Catalog No. N05841G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N05841G Supplier TCI America Supplier No. N05841G
Only null left

Chemical Identifiers

CAS 4533-42-0
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.186
MDL Number MFCD00119340
InChI Key PWCFKNYSCGRNRW-UHFFFAOYSA-N
PubChem CID 272431
IUPAC Name 1-(4-nitrophenyl)pyrrole
SMILES C1=CN(C=C1)C2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Color Yellow
Quantity 1 g
Formula Weight 188.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Nitrophenyl)pyrrole