missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Nitrophenyl)piperazine, 98%

Catalog No. p-7047321
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1870014 25 g
AAL1870022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1870014 Supplier Thermo Scientific Chemicals Supplier No. L1870014
Only null left

CAS: 6269-89-2 | C10H13N3O2 | 207.233 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6269-89-2
Molecular Formula C10H13N3O2
Molecular Weight (g/mol) 207.233
MDL Number MFCD00005961
InChI Key VWOJSRICSKDKAW-UHFFFAOYSA-N
Synonym 1-4-nitrophenyl piperazine, 1-4-nitro-phenyl-piperazine, piperazine, 1-4-nitrophenyl, 1-4-nitrophenyl-piperazine, n-4-nitrophenyl piperazine, n-4-nitrophenyl-piperazine, 4-nitrophenyl piperazine, 1-4-nitrophenyl piperazin, pubchem8580, 4-nitrophenylpiperazine
PubChem CID 80447
SMILES C1CN(CCN1)C2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 129°C to 133°C
Quantity 25 g
UN Number UN2811
Formula Weight 207.23
Percent Purity ≥98%
Chemical Name or Material 1-(4-Nitrophenyl)piperazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 228-443-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only