missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Nitrophenyl)-3-methyl-5-pyrazolone 98.0+%, TCI America™

Catalog No. M037725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M037725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M037725G Supplier TCI America Supplier No. M037725G
Only null left

Chemical Identifiers

CAS 6402-09-1
Molecular Formula C10H9N3O3
Molecular Weight (g/mol) 219.20
MDL Number MFCD00020751
InChI Key MYPAMGFTHVEING-UHFFFAOYSA-N
Synonym 3-Methyl-1-(4-nitrophenyl)-5-pyrazolone

Specifications

Melting Point 222°C
Color Yellow
Quantity 25 g
Formula Weight 219.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Nitrophenyl)-3-methyl-5-pyrazolone