missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Nitrophenyl)-3-methyl-5-phenylformazan 98.0+%, TCI America™

Catalog No. b0312100mg
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0312100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B0312100MG Supplier TCI America Supplier No. B0312100MG

Chemical Identifiers

CAS 68420-26-8
Molecular Formula C14H13N5O2
Molecular Weight (g/mol) 283.291
MDL Number MFCD00059829
InChI Key FHIGZTOTTOEVNT-DNTUAUKYSA-N
Synonym 5-Phenyl-1-(4-nitrophenyl)-3-methylformazan
PubChem CID 5819184
IUPAC Name N'-anilino-N-(4-nitrophenyl)iminoethanimidamide
SMILES CC(=NNC1=CC=CC=C1)N=NC2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 160°C
Quantity 100 mg
UN Number 1325
Formula Weight 283.29
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Nitrophenyl)-3-methyl-5-phenylformazan