missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Morpholinyl)-2-propanol, 98%

Catalog No. AAL1185909
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1185909 10 g
AAL1185918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1185909 Supplier Thermo Scientific Chemicals Supplier No. L1185909
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2109-66-2 | C7H15NO2 | 145.202 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2109-66-2
Molecular Formula C7H15NO2
Molecular Weight (g/mol) 145.202
MDL Number MFCD00023378
InChI Key YAXQOLYGKLGQKA-UHFFFAOYSA-N
Synonym n-2-hydroxypropyl morpholine, 1-morpholin-4-yl propan-2-ol, 1-morpholinopropan-2-ol, 1-morpholino-2-propanol, 4-2-hydroxypropyl morpholine, 1-4-morpholino-2-propanol, 4-morpholineethanol, alpha-methyl, 4-morpholineethanol, .alpha.-methyl, acmc-1ck5f, 1-morpholino propan-2-ol
PubChem CID 97957
IUPAC Name 1-morpholin-4-ylpropan-2-ol
SMILES CC(CN1CCOCC1)O

Specifications

Density 1.02
Boiling Point 218°C
Flash Point 92°C (197°F)
Refractive Index 1.4635
Quantity 10 g
Beilstein 1443
Formula Weight 145.2
Percent Purity 98%
Chemical Name or Material 1-(4-Morpholinyl)-2-propanol
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
EINECSNumber 218-298-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only