missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-Methoxyphenyl)piperazine Dihydrochloride 98.0+%, TCI America™
$53.68 - $150.98
Chemical Identifiers
| CAS | 38869-47-5 |
|---|---|
| Molecular Formula | C11H18Cl2N2O |
| Molecular Weight (g/mol) | 265.178 |
| MDL Number | MFCD00012767 |
| InChI Key | MMXANKLJIHSIQH-UHFFFAOYSA-N |
| Synonym | 1-4-methoxyphenyl piperazine dihydrochloride, 1-4-methoxyphenyl piperazinedihydrochloride, piperazine, 1-4-methoxyphenyl-, dihydrochloride, 1-4-methoxylphenyl-piperazine dihydrochloride, para-methoxyphenylpiperazine dihydrochloride, 1-4-methoxyphenyl-piperazine dihydrochloride, 4-methoxy-1-piperazinylbenzene, chloride, chloride, pubchem8579, acmc-1ct2c, ksc222g2n |
| PubChem CID | 2723700 |
| IUPAC Name | 1-(4-methoxyphenyl)piperazine;dihydrochloride |
| SMILES | COC1=CC=C(C=C1)N2CCNCC2.Cl.Cl |
Chemical Identifiers
| 38869-47-5 | |
| 265.178 | |
| MMXANKLJIHSIQH-UHFFFAOYSA-N | |
| 2723700 | |
| COC1=CC=C(C=C1)N2CCNCC2.Cl.Cl |
| C11H18Cl2N2O | |
| MFCD00012767 | |
| 1-4-methoxyphenyl piperazine dihydrochloride, 1-4-methoxyphenyl piperazinedihydrochloride, piperazine, 1-4-methoxyphenyl-, dihydrochloride, 1-4-methoxylphenyl-piperazine dihydrochloride, para-methoxyphenylpiperazine dihydrochloride, 1-4-methoxyphenyl-piperazine dihydrochloride, 4-methoxy-1-piperazinylbenzene, chloride, chloride, pubchem8579, acmc-1ct2c, ksc222g2n | |
| 1-(4-methoxyphenyl)piperazine;dihydrochloride |
Specifications
| 38869-47-5 | |
| C11H18Cl2N2O | |
| 5 g | |
| 1-4-methoxyphenyl piperazine dihydrochloride, 1-4-methoxyphenyl piperazinedihydrochloride, piperazine, 1-4-methoxyphenyl-, dihydrochloride, 1-4-methoxylphenyl-piperazine dihydrochloride, para-methoxyphenylpiperazine dihydrochloride, 1-4-methoxyphenyl-piperazine dihydrochloride, 4-methoxy-1-piperazinylbenzene, chloride, chloride, pubchem8579, acmc-1ct2c, ksc222g2n | |
| COC1=CC=C(C=C1)N2CCNCC2.Cl.Cl | |
| 265.178 | |
| 265.18 | |
| Crystalline Powder |
| Red-White | |
| MFCD00012767 | |
| 2811 | |
| MMXANKLJIHSIQH-UHFFFAOYSA-N | |
| 1-(4-methoxyphenyl)piperazine;dihydrochloride | |
| 2723700 | |
| ≥98.0% (HPLC,T) | |
| 1-(4-Methoxyphenyl)piperazine Dihydrochloride |
Safety and Handling
TSCA : No